Anti-B2M Mouse Monoclonal Antibody (CF640R) [clone: 246-E9.E7]

Supplier: Biotium

BNC401039-100 BNC401039-500
75883-918EA 325.12 CAD
75883-918 75883-920
Anti-B2M Mouse Monoclonal Antibody (CF640R) [clone: 246-E9.E7]
Antibodies
This antibody recognizes a protein of 12 kDa, identified asβ2-microglobulin.β2-microglobulin non-covalently associates with the 44 kDa chain to form the HLA Class I antigen complex. Human β2-microglobulin associated with HLA Class I antigens is expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes. This MAb is specific to human β2-microglobulin and does not react with non-human primate cells. This antibody reacts with all cell types excluding erythrocytes. The detection of β2-microglobulin in body fluids has been used as a tumor marker and for monitoring patients with HIV infection. Clone 246 E9.E7 is the same as HLA.ABC.m2.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: 246-E9.E7
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR