Anti-B2M Mouse Monoclonal Antibody (CF405S) [clone: 246-E9.E7]

Supplier: Biotium

BNC041039-100 BNC041039-500
75881-986EA 325.12 CAD
75881-986 75881-988
Anti-B2M Mouse Monoclonal Antibody (CF405S) [clone: 246-E9.E7]
Antibodies
This antibody recognizes a protein of 12 kDa, identified asβ2-microglobulin.β2-microglobulin non-covalently associates with the 44 kDa chain to form the HLA Class I antigen complex. Human β2-microglobulin associated with HLA Class I antigens is expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes. This MAb is specific to human β2-microglobulin and does not react with non-human primate cells. This antibody reacts with all cell types excluding erythrocytes. The detection of β2-microglobulin in body fluids has been used as a tumor marker and for monitoring patients with HIV infection. Clone 246 E9.E7 is the same as HLA.ABC.m2.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: 246-E9.E7
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR