Anti-ITGA2B Mouse Monoclonal Antibody (Biotin) [clone: ITGA2B/1036]

Supplier: Biotium

BNCB1036-100 BNCB1036-500
75895-694EA 325.12 CAD
75895-694 75895-696
Anti-ITGA2B Mouse Monoclonal Antibody (Biotin) [clone: ITGA2B/1036]
Antibodies
This antibody recognizes a calcium-dependent complex of CD41/CD61, a dimer of 90 kDa and 140 kDa present on the membrane of normal platelets and megakaryocytes. CD41/CD61 is also known as platelet glycoprotein GPIIb/GPIIIa or integrin IIa/3. This complex is the receptor of fibrinogen, fibronectin and von Willebrand factor, and mediates platelet adhesion and aggregation.

Type: Primary
Antigen: ITGA2B
Clonality: Monoclonal
Clone: Clone ITGA2B/1036
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR