Anti-ITGA2B Mouse Monoclonal Antibody (CF405S) [clone: ITGA2B/1036]

Supplier: Biotium

BNC041036-100 BNC041036-500
75895-742EA 325.12 CAD
75895-742 75895-744
Anti-ITGA2B Mouse Monoclonal Antibody (CF405S) [clone: ITGA2B/1036]
Antibodies
This antibody recognizes a calcium-dependent complex of CD41/CD61, a dimer of 90 kDa and 140 kDa present on the membrane of normal platelets and megakaryocytes. CD41/CD61 is also known as platelet glycoprotein GPIIb/GPIIIa or integrin IIa/3. This complex is the receptor of fibrinogen, fibronectin and von Willebrand factor, and mediates platelet adhesion and aggregation.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: ITGA2B
Clonality: Monoclonal
Clone: Clone ITGA2B/1036
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR