Anti-CD47 Mouse Monoclonal Antibody (Biotin) [clone: IAP/964 B6H12.2]

Supplier: Biotium

BNCB1029-100 BNCB1029-500
75913-842EA 325.12 CAD
75913-842 75913-844
Anti-CD47 Mouse Monoclonal Antibody (Biotin) [clone: IAP/964 B6H12.2]
Antibodies
This antibody reacts with Ig domain of CD47 protein. CD47, originally named integrin-associated protein (IAP), is a 50 kDa protein containing five membrane-spanning sequences and a short cytoplasmic tail. CD47 plays a role in both cell adhesion by acting as an adhesion receptor for THBS1 on platelets, and in the modulation of integrins. It is important in memory formation and synaptic plasticity in the hippocampus. CD47 may play a role in membrane transport and/or integrin dependent signal transduction.

Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: IAP/964 B6H12.2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR