Anti-CD70 Mouse Monoclonal Antibody (CF405S) [clone: TNFS7/1026]
Supplier: Biotium
BNC041026-100 BNC041026-500
75918-886EA 325.12 CAD
75918-886 75918-888
Anti-CD70 Mouse Monoclonal Antibody (CF405S) [clone: TNFS7/1026]
Antibodies
This antibody recognizes a protein of 30 kDa, identified as CD70. It is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T- and B-lymphocytes. It induces proliferation of co-stimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis.
CF® dyes are Biotium's next-generationMore Product Information