Anti-ICAM3 Mouse Monoclonal Antibody (Biotin) [clone: ICAM3/1019]

Supplier: Biotium

BNCB1019-100 BNCB1019-500
75912-044EA 325.12 CAD
75912-044 75912-046
Anti-ICAM3 Mouse Monoclonal Antibody (Biotin) [clone: ICAM3/1019]
Antibodies
This antibody recognizes an N-glycosylated glycoprotein of 120 kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3. CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signaling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This MAb is excellent for staining of formalin/paraffin tissues.

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: Clone ICAM3/1019
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR