Anti-ICAM3 Mouse Monoclonal Antibody (CF488A) [clone: ICAM3/1019]

Supplier: Biotium

BNC881019-100 BNC881019-500
75914-254EA 325.12 CAD
75914-254 75914-256
Anti-ICAM3 Mouse Monoclonal Antibody (CF488A) [clone: ICAM3/1019]
Antibodies
This antibody recognizes an N-glycosylated glycoprotein of 120 kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3. CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signaling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This MAb is excellent for staining of formalin/paraffin tissues.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®488A is a green fluorescent dye (Ex/Em 490/515 nm) with excellent brightness and photostability. The dye is minimally charged for less non-specific binding. CF®488A also is compatible with super-resolution imaging by TIRF.

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: Clone ICAM3/1019
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR