Anti-ACTA1 (Skeletal); ACTA2 (Smooth); ACTG2 (Smooth) Mouse Monoclonal Antibody (CF568) [clone: HHF35 MSA/953]

Supplier: Biotium

BNC681013-100 BNC681013-500
75968-276EA 325.12 CAD
75968-276 75968-278
Anti-ACTA1 (Skeletal); ACTA2 (Smooth); ACTG2 (Smooth) Mouse Monoclonal Antibody (CF568) [clone: HHF35 MSA/953]
Antibodies
This antibody recognizes actin of skeletal, cardiac, and smooth muscle cells. It is not reactive with other mesenchymal cells except for myoepithelium. Actin can be resolved on the basis of its isoelectric points into three distinctive components: alpha, beta, and gamma in order of increasing isoelectric point. Anti-muscle specific actin recognizes alpha and gamma isotypes of all muscle groups. Non-muscle cells such as vascular endothelial cells and connective tissues are non-reactive. Also, neoplastic cells of non-muscle-derived tissue such as carcinomas, melanomas, and lymphomas are negative.It stains tumors of smooth muscle (leiomyomas and leiomyosarcomas) as well as skeletal muscle (rhabdomyomas and rhabdomyosarcomas).

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: ACTA1 (Skeletal); ACTA2 (Smooth); ACTG2 (Smooth)
Clonality: Monoclonal
Clone: Clone HHF35 MSA/953
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR