Anti-CEACAM1 & CEACAM5 Mouse Monoclonal Antibody (Biotin) [clone: C66/1009]

Supplier: Biotium

BNCB1009-100 BNCB1009-500
75917-322EA 325.12 CAD
75917-322 75917-324
Anti-CEACAM1 & CEACAM5 Mouse Monoclonal Antibody (Biotin) [clone: C66/1009]
Antibodies
This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci of gastric carcinoma and in distinguishing pulmonary adenocarcinomas (60-70% are CEA positive) from pleural mesotheliomas (rarely or weakly CEA positive) . Anti-CEA positivity is seen in adenocarcinomas from the lung, colon, stomach, esophagus, pancreas, gallbadder, urachus, salivary gland, ovary, and endocervix.

Type: Primary
Antigen: CEACAM1 & CEACAM5
Clonality: Monoclonal
Clone: Clone C66/1009
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR