Anti-MUC5AC Mouse Monoclonal Antibody (Biotin) [clone: 58M1]

Supplier: Biotium

BNCB1003-100 BNCB1003-500
75962-550EA 325.12 CAD
75962-550 75962-552
Anti-MUC5AC Mouse Monoclonal Antibody (Biotin) [clone: 58M1]
Antibodies
This MAb recognizes the peptide core of gastric mucin M1 (recently identified as Mucin 5AC). Its epitope is destroyed by beta-mercaptoethanol but not by periodate treatment. This mucin is present in primary ovarian mucinous cancer but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. Together with a panel of antibodies, Anti-MUC5AC may be useful for differential identification of primary mucinous ovarian tumors from colon adenocarcinoma metastatic to the ovary. MUC5AC antibodies may also be useful for identification of intestinal metaplasia as well as in the identification of pancreatic carcinoma and pre-cancerous changes vs. normal pancreas.

Type: Primary
Antigen: MUC5AC
Clonality: Monoclonal
Clone: Clone 58M1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa s
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR