Anti-CD22 Mouse Monoclonal Antibody (CF488A) [clone: Myg13]

Supplier: Biotium

BNC880974-100 BNC880974-500
75891-086EA 325.12 CAD
75891-086 75891-088
Anti-CD22 Mouse Monoclonal Antibody (CF488A) [clone: Myg13]
Antibodies
This antibody recognizes a protein of 130-140 kDa, identified as CD22 (also known as BL-CAM). CD22 expression is restricted to normal and neoplastic B cells and is absent from other haemopoietic cell types. In B-cell ontogeny, CD22 is first expressed in the cytoplasm of pro-B and pre-B cells, and on the surface as B cells mature to become IgD . It is not expressed by plasma cells, CD22 is found highly expressed in follicular mantle and marginal zone B-cells, and while germinal center B-cells are relatively weak. CD22 is a member of the immunoglobulin superfamily and serves as an adhesionMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD22
Antigen name CD22
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 488A
Clone Myg13
Reactivity Human,Rat,Mouse
Host Mouse
Gene ID Unigene ID: 579691
Isotype IgG1
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR