Anti-MYADM Mouse Monoclonal Antibody (CF405S) [clone: MYADM/972]

Supplier: Biotium

BNC040972-100 BNC040972-500
75964-604EA 325.12 CAD
75964-604 75964-606
Anti-MYADM Mouse Monoclonal Antibody (CF405S) [clone: MYADM/972]
Antibodies
This antibody recognizes a myeloid associated differentiation antigen in the cytoplasm of mature granulocytes. It shows no reactivity with any other cell type in human tissues. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. It reacts with early precursor and mature forms of human and monkey myeloid cells. This MAb is useful for the detection of myeloid leukemias and granulocytic sarcomas. It can be used as a marker of granulocytes in normal tissues or inflammatory processes.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: MYADM
Clonality: Monoclonal
Clone: Clone MYADM/972
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Macaque
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR