Anti-CD63 Mouse Monoclonal Antibody [clone: LAMP3/968]

Supplier: Biotium

BNUB0968-100 BNUB0968-500
75916-658EA 380.65 CAD
75916-658 75916-660
Anti-CD63 Mouse Monoclonal Antibody [clone: LAMP3/968]
Antibodies
This MAb recognizes protein of 26 kDa-60 kDa, which is identified as CD63. Its epitope is different from that of MAb LAMP3/529. The tetraspanins are integral membrane proteins expressed on cell surface and granular membranes of hematopoietic cells and are components of multi-molecular complexes with specific integrins. The tetraspanin CD63 is a lysosomal membrane glycoprotein that translocates to the plasma membrane after platelet activation. CD63 is expressed on activated platelets, monocytes and macrophages, and is weakly expressed on granulocytes, T cell and B cells. It is located on the basophilic granule membranes and on the plasma membranes of lymphocytes and granulocytes. CD63 is a member of the TM4 superfamily of leukocyte glycoproteins that includes CD9, CD37 and CD53, which contain four transmembrane regions. CD63 may play a role in phagocytic and intracellular lysosome-phagosome fusion events. CD63 deficiency is associated with Hermansky-Pudlak syndrome and is strongly expressed during the early stages of melanoma progression.

Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: Clone LAMP3/968
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa (confirmed and correct)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR