Anti-BSG Mouse Monoclonal Antibody (CF640R) [clone: BSG/963]

Supplier: Biotium

BNC400963-500 BNC400963-100
75887-572EA 900.17 CAD
75887-572 75887-762
Anti-BSG Mouse Monoclonal Antibody (CF640R) [clone: BSG/963]
Antibodies
This MAb recognizes extracellular epitope of human CD147. It is expressed more intensely on thymocytes than on mature peripheral blood T cells. CD147 is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. It stimulates the production of interstitial collagenase, gelatinase A, stromelysin-1 and various metalloproteinases (MMPs) by fibroblasts. These enzymes are important factors in cancer invasion and metastasis.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: Clone BSG/963
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR