Anti-H1 Mouse Monoclonal Antibody [clone: HH1/957]

Supplier: Biotium

BNUB0957-100 BNUB0957-500
75950-672EA 380.65 CAD
75950-672 75950-674
Anti-H1 Mouse Monoclonal Antibody [clone: HH1/957]
Antibodies
Eukaryotic histones are basic and water-soluble nuclear proteins. They form hetero-octameric nucleosome particles by wrapping 146 base pairs of DNA in a left-handed super-helical turn sequentially to form chromosomal fiber. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form the octamer; formed of two H2A-H2B dimers and two H3-H4 dimers, forming two nearly symmetrical halves by tertiary structure. Over 80% of nucleosomes contain the linker Histone H1, derived from an intronless gene that interacts with linker DNA between nucleosomes and mediates compaction into higher order chromatin. Histones are subject to posttranslational modification by enzymes primarily on their N-terminal tails, but also in their globular domains. Such modifications include methylation, citrullination, acetylation, phosphorylation, sumoylation, ubiquitination and ADP-ribosylation.

Type: Primary
Antigen: H1
Clonality: Monoclonal
Clone: Clone HH1/957
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR