Anti-H1 Mouse Monoclonal Antibody (CF405S) [clone: HH1/957]

Supplier: Biotium

BNC040957-100 BNC040957-500
75952-952EA 325.12 CAD
75952-952 75952-954
Anti-H1 Mouse Monoclonal Antibody (CF405S) [clone: HH1/957]
Antibodies
Eukaryotic histones are basic and water-soluble nuclear proteins. They form hetero-octameric nucleosome particles by wrapping 146 base pairs of DNA in a left-handed super-helical turn sequentially to form chromosomal fiber. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form the octamer; formed of two H2A-H2B dimers and two H3-H4 dimers, forming two nearly symmetrical halves by tertiary structure. Over 80% of nucleosomes contain the linker Histone H1, derived from an intronless gene that interacts with linker DNA between nucleosomes and mediates compaction into higherMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol H1
Environmentally Preferable
Antigen name Histone H1
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 405S
Clone HH1/957
Reactivity Human,Rat,Mouse
Host Mouse
Gene ID Unigene ID: 226117 , 97358
Isotype IgG2a kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR