Anti-UACA Mouse Monoclonal Antibody (Biotin) [clone: AE-5]
Supplier: Biotium
BNCB0956-100 BNCB0956-500
75978-768EA 325.12 CAD
75978-768 75978-770
Anti-UACA Mouse Monoclonal Antibody (Biotin) [clone: AE-5]
Antibodies
UACA (Uveal Autoantigen with Coiled-coil domains and Ankyrin repeats) is a 1,416 amino acid nuclear membrane protein. It was originally identified as an autoantigen in patients with panuveitis, a characteristic of Vogt-Koyanagi-Harada disease, and in patients with Graves' disease. UACA was also later identified as Nucling, an mRNA differentially expressed in F9 embryonal carcinoma cells during cardiac muscle differentiation. UACA appears to function as a pro-apoptotic protein that recruits the apaf-1-pro-caspase-9 complex for the induction of apoptosis to mediate the cell-death pathway.