Anti-IGHG Mouse Monoclonal Antibody [clone: B33/20]

Supplier: Biotium

BNUB0952-100 BNUB0952-500
75954-114EA 380.65 CAD
75954-114 75954-116
Anti-IGHG Mouse Monoclonal Antibody [clone: B33/20]
Antibodies
This antibody recognizes a protein of 75 kDa, identified as γ heavy chain of human immunoglobulins. Its epitope maps in CH2 domain of Fc region of IgG. It reacts with all sub-classes of γ chain of human immunoglobulins. It does not cross-react with α (IgA), μ (IgM), ε (IgE), or δ (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This MAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Type: Primary
Antigen: IGHG
Clonality: Monoclonal
Clone: Clone B33/20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR