Anti-KRT5 & KRT8 Mouse Monoclonal Antibody (CF594) [clone: C-50]

Supplier: Biotium

BNC940948-100 BNC940948-500
75919-924EA 325.12 CAD
75919-924 75919-926
Anti-KRT5 & KRT8 Mouse Monoclonal Antibody (CF594) [clone: C-50]
Antibodies
This antibody recognizes keratin 5 (58 kDa) and keratin 8 (52.5 kDa). Simple epithelia express cytokeratin 8 in combination with 18. On the other hand, basal cells of stratified epithelia express cytokeratin 5 paired with 14. This antibody therefore, reacts with a wide range of epithelia and their carcinomas.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®594 is a deep red fluorescent dye (Ex/Em 593/614 nm). It yields the brightest conjugates among spectrally similar dyes, and has excellent photostability.

Type: Primary
Antigen: KRT5 & KRT8
Clonality: Monoclonal
Clone: Clone C-50
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cow, Dog, Ferret, Hamster, Human, Mouse, Pig, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR