Anti-ABO Mouse Monoclonal Antibody [clone: 19-OLE]

Supplier: Biotium

BNUB0947-100 BNUB0947-500
75888-800EA 380.65 CAD
75888-800 75888-802
Anti-ABO Mouse Monoclonal Antibody [clone: 19-OLE]
Antibodies
This antibody recognizes the blood group H type 2 antigens, trisaccharide Fuca1-2Galb1-4GlcNAc of human origin. This protein is the basis of the ABO blood group system. The histo-blood group ABO involves three carbohydrate antigens: A, B, and H. A, B, and AB individuals express a glycosyltransferase activity that converts the H antigen to the A antigen (by addition of UDP-GalNAc) or to the B antigen (by addition of UDP-Gal), whereas O individuals lack such activity. It is expressed on endothelial cells, epithelial cells and granulocytes. Increased expression of this antigen has been observed on some tumor tissues such as gastric carcinomas, urothelial carcinomas, and colon carcinomas.

Type: Primary
Antigen: ABO
Clonality: Monoclonal
Clone: Clone 19-OLE
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR