Anti-Double Stranded DNA Mouse Monoclonal Antibody (CF594) [clone: AE-2]

Supplier: Biotium

BNC940946-100 BNC940946-500
75924-248EA 325.12 CAD
75924-248 75924-250
Anti-Double Stranded DNA Mouse Monoclonal Antibody (CF594) [clone: AE-2]
Antibodies
This monoclonal antibody is part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. This MAb recognizes the double stranded DNA in human cells. It can be used to stain the nuclei in cell or tissue preparations and can be used as a nuclear marker in human cells. This MAb produces a homogeneous staining pattern in the nucleus of normal and malignant cells.,Double Stranded deoxyribonucleic acid (ds DNA) is the genetic material of allMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name Double Stranded DNA
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 594
Clone AE-2
Reactivity Human
Host Mouse
Isotype IgG3 kappa
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR