Anti-MFGE8 Mouse Monoclonal Antibody (CF488A) [clone: EDM45]

Supplier: Biotium

BNC880942-100 BNC880942-500
75961-270EA 325.12 CAD
75961-270 75961-272
Anti-MFGE8 Mouse Monoclonal Antibody (CF488A) [clone: EDM45]
Antibodies
This antibody recognizes a protein of 40-45 kDa, identified as human milk fat globule membrane protein (HMFG). HMFG is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as to the outer surface of the human milk fat globule. HMFG is considered as a differentiation marker. It is useful as specific breast epithelial marker and can also provide a tool to study the role of the cell surface in normal and neoplastic mammary development.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®488A is a green fluorescent dye (Ex/Em 490/515 nm) with excellent brightness and photostability. The dye is minimally charged for less non-specific binding. CF®488A also is compatible with super-resolution imaging by TIRF.

Type: Primary
Antigen: MFGE8
Clonality: Monoclonal
Clone: Clone EDM45
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR