Anti-HSP90B1 Rat Monoclonal Antibody (CF405S) [clone: Clone 9G10.F8.2]

Supplier: Biotium

BNC040940-100 BNC040940-500
75949-346EA 325.12 CAD
75949-346 75949-348
Anti-HSP90B1 Rat Monoclonal Antibody (CF405S) [clone: Clone 9G10.F8.2]
Antibodies
This antibody recognizes a protein of 94 kDa, which is identified as the glucose-regulated protein 94 (grp94) and also tumor rejection antigen (gp96). Grp94 shows a high degree of sequence homology with the heat shock protein 90 (hsp90). This MAb is highly specific to grp94 and shows minimal cross-reaction with other members of the HSP90 family. Grp s are a class of proteins unresponsive to heat shock and are induced by glucose deprivation. Grp94 has been briefly studied as a prognostic factor in breast cancer.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: Clone 9G10.F8.2
Conjugation: CF405S
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Chicken, Cow, Dog, Guinea Pig, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus laevis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR