Anti-CD46 Mouse Monoclonal Antibody (CF405S) [clone: 169-1-E4.3]

Supplier: Biotium

BNC040930-100 BNC040930-500
75913-594EA 325.12 CAD
75913-594 75913-596
Anti-CD46 Mouse Monoclonal Antibody (CF405S) [clone: 169-1-E4.3]
Antibodies
This antibody recognizes a protein of 52 kDa-58 kDa, identified as CD46 (also known as membrane cofactor protein, MCP). Its epitope is located on the four immunogenic short consensus repeat (SCR) domains at N-terminus of the protein. CD46 exists as many isoforms in a variety of tissues. It is strongly expressed on salivary gland ducts and kidney ducts, moderately on lymphocytes and endothelium, and weakly on interstitial tissues and muscle cells, but not on erythrocytes. CD46 functions as a C3b/C4b-binding glycoprotein that inhibits complement activation on host cells. It also serves as a measles virus receptor, an adherence factor for group A Streptococcus pyogenes, and a cellular pilus receptor for pathogenic Neisseria.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: 169-1-E4.3
Conjugation: CF405S
Epitope:
Host: Mouse
Reactivity: Baboon, Human
Isotype: IgG2a, kappa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR