Anti-CD14 Mouse Monoclonal Antibody (CF640R) [clone: MDA/929]

Supplier: Biotium

BNC400929-100 BNC400929-500
75887-518EA 325.12 CAD
75887-518 75887-520
Anti-CD14 Mouse Monoclonal Antibody (CF640R) [clone: MDA/929]
Antibodies
This antibody recognizes a protein of 55 kDa, identified as CD14 (also known lipopolysaccharide receptor). CD14 is expressed strongly on monocytes and macrophage and weakly on the surface of neutrophils. CD14 is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a high affinity receptor for complexes of LPS and LPS binding protein (LBP). Soluble CD14, also binding to LPS, acts at physiological concentration as an LPS agonist and has, at higher concentrations, an LPS antagonizing effect in cell activation.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: MDA/929
Conjugation: CF640R
Epitope:
Host: Mouse
Reactivity: Human
Isotype: IgG1, kappa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR