Anti-FCGR3A Mouse Monoclonal Antibody (CF640R) [clone: HuNK2]

Supplier: Biotium

BNC400928-100 BNC400928-500
75887-930EA 325.12 CAD
75887-930 75887-932
Anti-FCGR3A Mouse Monoclonal Antibody (CF640R) [clone: HuNK2]
Antibodies
This antibody recognizes a protein of 50-65 kDa, identified as CD16 (Workshop IV; Code N39 ) (also known low affinity Fc receptor III for IgG (FcRIII) or Leu 11). CD16 exists as a polypepetide-anchored from (FCRIIIA or CD16A) on human natural killer (NK) cells and monocytes/ macrophages and as a glycosylphosphatidylinositol (GPI)-anchored form (FcRIIIB or CD16B) on neutrophils. CD16B is polymorphic and the two alleles are termed NA1 and NA2.3 CD16 plays a role in signal transduction, NK cell activation and antibody-dependent cellular cytotoxicity. This MAb has been showed to inhibit the binding of immune complex to NK cells, inhibit cytotoxicity of NK cells, and induce calcium fluxes in NK cells and neutrophils.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: FCGR3A
Clonality: Monoclonal
Clone: HuNK2
Conjugation: CF640R
Epitope:
Host: Mouse
Reactivity: Human
Isotype: IgG2a, kappa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR