Anti-CD44 Mouse Monoclonal Antibody (CF568) [clone: HCAM/918]

Supplier: Biotium

BNC680918-100 BNC680918-500
75912-586EA 325.12 CAD
75912-586 75912-588
Anti-CD44 Mouse Monoclonal Antibody (CF568) [clone: HCAM/918]
Antibodies
This antibody recognizes a cell surface glycoprotein of 80-95 kDa (CD44) on lymphocytes, monocytes, and granulocytes (Leucocyte Typing Workshop V). Its epitope is resistant to digestion by trypsin and chymotrypsin. The CD44 family of glycoproteins exists in a number of variant isoforms, the most common being the standard 85-95 kDa or hematopoietic variant (CD44s). Higher molecular weight isoforms are described in epithelial cells (CD44v), which are believed to function in intercellular adhesion and stromal binding. CD44 immunostaining is commonly used for the discrimination of urothelial transitional cell carcinoma in-situ from non-neoplastic changes in the urothelium.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: HCAM/918
Conjugation: CF568
Epitope:
Host: Mouse
Reactivity: Human, Non-human primates
Isotype: IgG2a, kappa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR