Anti-CD44 Mouse Monoclonal Antibody (CF647) [clone: HCAM/918]

Supplier: Biotium

BNC470918-100 BNC470918-500
75911-316EA 325.12 CAD
75911-316 75911-318
Anti-CD44 Mouse Monoclonal Antibody (CF647) [clone: HCAM/918]
Antibodies
This antibody recognizes a cell surface glycoprotein of 80-95 kDa (CD44) on lymphocytes, monocytes, and granulocytes (Leucocyte Typing Workshop V). Its epitope is resistant to digestion by trypsin and chymotrypsin. The CD44 family of glycoproteins exists in a number of variant isoforms, the most common being the standard 85-95 kDa or hematopoietic variant (CD44s). Higher molecular weight isoforms are described in epithelial cells (CD44v), which are believed to function in intercellular adhesion and stromal binding. CD44 immunostaining is commonly used for the discrimination of urothelial transitional cell carcinoma in-situ from non-neoplastic changes in the urothelium.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®647 is a far-red fluorescent dye (Ex/Em 650/665 nm) with excellent brightness. It also is compatible with super-resolution imaging by STORM.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: HCAM/918
Conjugation: CF647
Epitope:
Host: Mouse
Reactivity: Human, Non-human primates
Isotype: IgG2a, kappa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR