Anti-CD44 Mouse Monoclonal Antibody (CF640R) [clone: HCAM/918]

Supplier: Biotium

BNC400918-100 BNC400918-500
75911-312EA 325.12 CAD
75911-312 75911-314
Anti-CD44 Mouse Monoclonal Antibody (CF640R) [clone: HCAM/918]
Antibodies
This antibody recognizes a cell surface glycoprotein of 80-95 kDa (CD44) on lymphocytes, monocytes, and granulocytes (Leucocyte Typing Workshop V). Its epitope is resistant to digestion by trypsin and chymotrypsin. The CD44 family of glycoproteins exists in a number of variant isoforms, the most common being the standard 85-95 kDa or hematopoietic variant (CD44s). Higher molecular weight isoforms are described in epithelial cells (CD44v), which are believed to function in intercellular adhesion and stromal binding. CD44 immunostaining is commonly used for the discrimination of urothelialMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD44
Antigen name CD44 Standard
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 640R
Clone HCAM/918
Reactivity Non-human primates,Human
Host Mouse
Gene ID Unigene ID: 502328
Isotype IgG2a kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR