Anti-PTH Mouse Monoclonal Antibody (CF488A) [clone: PTH/911]

Supplier: Biotium

BNC880911-100 BNC880911-500
75972-860EA 325.12 CAD
75972-860 75972-862
Anti-PTH Mouse Monoclonal Antibody (CF488A) [clone: PTH/911]
Antibodies
The epitope of this MAb maps in the N-terminus of PTH, a hormone produced by the parathyroid gland that regulates the concentration of calcium and phosphorus in extracellular fluid. This hormone elevates blood Ca2 levels by dissolving the salts in bone and preventing their renal excretion.It is produced in the parathyroid gland as an 84 amino acid single chain polypeptide. It can also be secreted as N-terminal truncated fragments or C-terminal fragments after intracellular degradation, as in case of hypercalcemia. Defects in this gene are a cause of familial isolated hypoparathyroidism (FIH);More Product Information
Order Now

SPECIFICATIONS

Antigen symbol PTH
Environmentally Preferable
Antigen name PTH / Parathyroid Hormone
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 488A
Clone PTH/911
Reactivity Human
Host Mouse
Gene ID Unigene ID: 37045
Isotype IgG2b kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR