Anti-DPP4 Mouse Monoclonal Antibody (CF568) [clone: DPP4/910]

Supplier: Biotium

BNC680910-100 BNC680910-500
75891-534EA 325.12 CAD
75891-534 75891-536
Anti-DPP4 Mouse Monoclonal Antibody (CF568) [clone: DPP4/910]
Antibodies
This antibody recognizes a glycoprotein of 110 kDa, identified as CD26. It is an atypical serine protease belonging to the prolyl oligopeptidase family. It is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: DPP4
Clonality: Monoclonal
Clone: Clone DPP4/910
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR