Anti-DPP4 Mouse Monoclonal Antibody (CF660R) [clone: DPP4/910]

Supplier: Biotium

BNC610910-100 BNC610910-500
75891-550EA 325.12 CAD
75891-550 75891-552
Anti-DPP4 Mouse Monoclonal Antibody (CF660R) [clone: DPP4/910]
Antibodies
This antibody recognizes a glycoprotein of 110 kDa, identified as CD26. It is an atypical serine protease belonging to the prolyl oligopeptidase family. It is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®660R is a far-redMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol DPP4
Antigen name CD26
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 660R
Clone DPP4/910
Reactivity Human,Rat
Host Mouse
Gene ID Unigene ID: 368912
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR