Anti-TYR Mouse Monoclonal Antibody [clone: T311 OCA1/812]

Supplier: Biotium

BNUB0908-100 BNUB0908-500
75978-538EA 380.65 CAD
75978-538 75978-540
Anti-TYR Mouse Monoclonal Antibody [clone: T311 OCA1/812]
Antibodies
This antibody recognizes a cluster of proteins between 70-80 kDa, identified as tyrosinase. Occasionally a minor band at 55 kDa is also detected. This MAb shows no cross-reaction with MAGE-1 and tyrosinase-related protein 1, TRP-1/gp75. Tyrosinase is a copper-containing metalloglycoprotein that catalyzes several steps in the melanin pigment biosynthetic pathway; the hydroxylation of tyrosine to L-3,4-dihydroxy-phenylalanine (dopa), and the subsequent oxidation of dopa to dopaquinone. Mutations of the tyrosinase gene occur in various forms of albinism. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients. Staining of melanomas with this MAb shows tyrosinase in melanotic as well as amelanotic variants. This MAb is a useful marker for melanocytes and melanomas.

Type: Primary
Antigen: TYR
Clonality: Monoclonal
Clone: Clone T311 OCA1/812
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG's
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR