Anti-NKX2-1 Mouse Monoclonal Antibody (CF640R) [clone: 8G7G3/1 NX2.1/690]

Supplier: Biotium

BNC400907-100 BNC400907-500
75978-192EA 325.12 CAD
75978-192 75978-194
Anti-NKX2-1 Mouse Monoclonal Antibody (CF640R) [clone: 8G7G3/1 NX2.1/690]
Antibodies
This antibody recognizes a protein of 40 kDa, identified as Thyroid transcription factor-1 (TTF-1). TTF-1 is a member of the NKx2 family of homeodomain transcription factors. It is expressed in epithelial cells of the thyroid gland and the lung. Nuclei from liver, stomach, pancreas, small intestine, colon, kidney, breast, skin, testes, pituitary, prostate, and adrenal glands are unreactive. Anti-TTF-1 is useful in differentiating primary adenocarcinoma of the lung from metastatic carcinomas originating in the breast, mediastinal germ cell tumors, and malignant mesothelioma. It can also be used to differentiate small cell lung carcinoma from lymphoid infiltrates.Loss of TTF-1 expression in non-small cell lung carcinoma has been associated with aggressive behavior of such neoplasms. TTF-1 reactivity is also seen in thyroid malignancies.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: NKX2-1
Clonality: Monoclonal
Clone: Clone 8G7G3/1 NX2.1/690
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG's
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR