Anti-Mitochondrial Marker Mouse Monoclonal Antibody [clone: AE-1]

Supplier: Biotium

BNUB0905-100 BNUB0905-500
75961-704EA 380.65 CAD
75961-704 75961-706
Anti-Mitochondrial Marker Mouse Monoclonal Antibody [clone: AE-1]
Antibodies
This MAb recognizes a 60 kDa antigen associated with the mitochondria in human cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the mitochondria in human cells only. It can be used to stain the mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-like pattern in normal and malignantMore Product Information
Order Now

SPECIFICATIONS

Antigen name Mitochondrial Marker
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone AE-1
Reactivity Human
Host Mouse
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.2 mg/mL
Storage buffer PBS/0.05% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR