Anti-CD34 Mouse Monoclonal Antibody (CF640R) [clone: QBEnd/10 HPCA1/763]

Supplier: Biotium

BNC400904-100 BNC400904-500
75894-682EA 325.12 CAD
75894-682 75894-684
Anti-CD34 Mouse Monoclonal Antibody (CF640R) [clone: QBEnd/10 HPCA1/763]
Antibodies
This MAb recognizes a single chain, transmembrane, heavily glycosylated protein of 90-120 kDa, which is identified as CD34. Its expression is a hallmark for identifying pluripotent hematopoietic stem or progenitor cells. Its expression is gradually lost as lineage committed progenitors differentiate. CD34 is a marker of choice for staining blasts in acute myeloid leukemia. In addition, CD34 is expressed by soft tissue tumors, such as solitary fibrous tumor and gastrointestinal stromal tumor. Its expression is also found in vascular endothelium. Additionally, it appears that proliferatingMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD34
Environmentally Preferable
Antigen name CD34
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 640R
Clone QBEnd/10 HPCA1/763
Reactivity Human
Host Mouse
Gene ID Unigene ID: 374990
Isotype IgGs
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR