Anti-CD34 Mouse Monoclonal Antibody (CF405S) [clone: QBEnd/10 HPCA1/763]

Supplier: Biotium

BNC040904-100 BNC040904-500
75894-658EA 325.12 CAD
75894-658 75894-660
Anti-CD34 Mouse Monoclonal Antibody (CF405S) [clone: QBEnd/10 HPCA1/763]
Antibodies
This MAb recognizes a single chain, transmembrane, heavily glycosylated protein of 90-120 kDa, which is identified as CD34. Its expression is a hallmark for identifying pluripotent hematopoietic stem or progenitor cells. Its expression is gradually lost as lineage committed progenitors differentiate. CD34 is a marker of choice for staining blasts in acute myeloid leukemia. In addition, CD34 is expressed by soft tissue tumors, such as solitary fibrous tumor and gastrointestinal stromal tumor. Its expression is also found in vascular endothelium. Additionally, it appears that proliferating endothelial cells express this molecule more than the non-proliferating endothelial cells. Anti-CD34 labels > 85% of angiosarcoma and Kaposi's sarcoma, but with a lower specificity.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: QBEnd/10 HPCA1/763
Conjugation: CF405S
Epitope:
Host: Mouse
Reactivity: Human
Isotype: IgGs
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR