Anti-KRT7 Mouse Monoclonal Antibody (CF647) [clone: KRT7/903]

Supplier: Biotium

BNC470903-100 BNC470903-500
75922-642EA 325.12 CAD
75922-642 75922-644
Anti-KRT7 Mouse Monoclonal Antibody (CF647) [clone: KRT7/903]
Antibodies
Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Antibody to cytokeratin is useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).

CF® dyes are Biotium's next-generation fluorescent dyes. CF®647 is a far-red fluorescent dye (Ex/Em 650/665 nm) with excellent brightness. It also is compatible with super-resolution imaging by STORM.

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: Clone KRT7/903
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR