Anti-KRT7 Mouse Monoclonal Antibody (CF640R) [clone: KRT7/903]

Supplier: Biotium

BNC400903-100 BNC400903-500
75920-316EA 325.12 CAD
75920-316 75920-318
Anti-KRT7 Mouse Monoclonal Antibody (CF640R) [clone: KRT7/903]
Antibodies
Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. Antibody to cytokeratin is useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: Clone KRT7/903
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR