Anti-NCL Mouse Monoclonal Antibody (Biotin) [clone: NCL/902]

Supplier: Biotium

BNCB0902-100 BNCB0902-500
75966-532EA 325.12 CAD
75966-532 75966-534
Anti-NCL Mouse Monoclonal Antibody (Biotin) [clone: NCL/902]
Antibodies
This antibody recognizes a protein of ~76 kDa, which is identified as Nucleolin (NCL). It is the major nucleolar phosphoprotein of growing eukaryotic cells. NCL is located mainly in dense fibrillar regions of the nucleolus. It is found associated with intranucleolar chromatin and pre-ribosomal particles. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. It induces chromatin decondensation by binding to histone H1. It is thought to play a role in pre-rRNA transcription and ribosome assembly.This MAb can be used to stain the nucleoli in cell or tissue preparations and can be used as a marker of the nucleoli in subcellular fractions. It produces a speckled pattern in the nuclei of cells of normal and malignant cells and may be used to stain the nucleoli of cells in fixed or frozen tissue sections. It can be used with paraformaldehyde fixed frozen tissue or cell preparations and formalin fixed, paraffin-embedded tissue sections.

Type: Primary
Antigen: NCL
Clonality: Monoclonal
Clone: NCL/902
Conjugation: Biotin
Epitope:
Host: Mouse
Reactivity: Human
Isotype: IgG1, kappa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR