Anti-NCL Mouse Monoclonal Antibody (CF640R) [clone: NCL/902]

Supplier: Biotium

BNC400902-100 BNC400902-500
75966-564EA 325.12 CAD
75966-564 75966-566
Anti-NCL Mouse Monoclonal Antibody (CF640R) [clone: NCL/902]
Antibodies
This antibody recognizes a protein of ~76 kDa, which is identified as Nucleolin (NCL). It is the major nucleolar phosphoprotein of growing eukaryotic cells. NCL is located mainly in dense fibrillar regions of the nucleolus. It is found associated with intranucleolar chromatin and pre-ribosomal particles. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. It induces chromatin decondensation by binding to histone H1. It is thought to play a role in pre-rRNA transcription and ribosome assembly.This MAb can be used to stain the nucleoli in cell or tissueMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol NCL
Antigen name Nucleolin
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 640R
Clone NCL/902
Reactivity Human
Host Mouse
Gene ID Unigene ID: 79110
Isotype IgG1 kappa
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR