Anti-MUC5AC Mouse Monoclonal Antibody (CF488A) [clone: CLH2]

Supplier: Biotium

BNC880897-500 BNC880897-100
75962-632EA 900.17 CAD
75962-632 75963-238
Anti-MUC5AC Mouse Monoclonal Antibody (CF488A) [clone: CLH2]
Antibodies
Mucin 5AC glycoprotein (MUC5AC) is a HMW glycoprotein belonging to the superfamily of mucins. Mucins are produced by epithelial cells and can be divided into two families; secretory mucins and membrane bound mucins. MUC5AC is a mucus-forming, secreted mucin that is found in normal gastric and tracheo-bronchial mucosa, but absent from normal colon. MUC5AC expression is present in primary ovarian mucinous cancer but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. Together with a panel of antibodies, Anti-MUC5AC may be useful for differentialMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol MUC5AC
Environmentally Preferable
Antigen name Mucin 5AC
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 488A
Clone CLH2
Reactivity Human
Host Mouse
Gene ID Unigene ID: 534332
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR