Anti-MUC6 Mouse Monoclonal Antibody (CF594) [clone: CLH5]

Supplier: Biotium

BNC940891-100 BNC940891-500
75962-874EA 325.12 CAD
75962-874 75962-876
Anti-MUC6 Mouse Monoclonal Antibody (CF594) [clone: CLH5]
Antibodies
The MUC6 gastric mucin is a secreted glycoprotein that plays an essential role in epithelial cyto-protection from acid, proteases, pathogenic microorganisms, and mechanical trauma in the gastrointestinal tract. Mucin 6 expression is highest in the stomach and gall bladder, with lower expression in the terminal ileum and right colon. In gastric cancer, Mucin 6 has an altered expression. In normal stomach, Mucin 6 is associated with Lewis type 2; Mucin 6 is also expressed in gastric metaplasia, duodenum and pancreas. Mucin 6 is a secretory mucin, located in the deeper mucosal folds of humanMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol MUC6
Environmentally Preferable
Antigen name Mucin 6
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 594
Clone CLH5
Reactivity Human
Host Mouse
Gene ID Unigene ID: 528432
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR