Anti-CYCS Mouse Monoclonal Antibody [clone: CTC05]

Supplier: Biotium

BNUB0887-100 BNUB0887-500
75923-810EA 380.65 CAD
75923-810 75923-812
Anti-CYCS Mouse Monoclonal Antibody [clone: CTC05]
Antibodies
Cytochrome c is a well-characterized mobile electron transport protein that is essential to energy conversion in all aerobic organisms. In mammalian cells, this highly conserved protein is normally localized to the mitochondrial inter-membrane space. More recent studies have identified cytosolic Cytochrome c as a factor necessary for activation of apoptosis. During apoptosis, Cytochrome c is trans-located from the mitochondrial membrane to the cytosol, where it is required for activation of caspase-3 (CPP32). Overexpression of Bcl-2 has been shown to prevent the translocation of Cytochrome c, thereby blocking the apoptotic process. Overexpression of Bax has been shown to induce the release of Cytochrome c and to induce cell death. The release of Cytochrome c from the mitochondria is thought to trigger an apoptotic cascade, whereby Apaf-1 binds to Apaf-3 (caspase-9) in a Cytochrome c-dependent manner, leading to caspase-9 cleavage of caspase-3.

Type: Primary
Antigen: CYCS
Clonality: Monoclonal
Clone: CTC05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Reactivity: Dog, Drosophila, Frog, Horse, Human, Mouse, Pigeon, Rat
Isotype: IgG2b, kappa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR