Anti-KLK3 Mouse Monoclonal Antibody (CF568) [clone: A67-B/E3]

Supplier: Biotium

BNC680883-100 BNC680883-500
75972-244EA 325.12 CAD
75972-244 75972-246
Anti-KLK3 Mouse Monoclonal Antibody (CF568) [clone: A67-B/E3]
Antibodies
This antibody recognizes a single protein of 33-34 kDa, identified as the prostate specific antigen (PSA). This MAb is highly specific to PSA and stains prostatic secretory and ductal epithelium in both normal and neoplastic tissues. PSA is a chymotrypsin-like serine protease (kallikrein family) exclusively produced by the prostate epithelium, and abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA isMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol KLK3
Antigen name PSA
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Clone A67-B/E3
Reactivity Human
Host Mouse
Gene ID Unigene ID: 171995
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR