Anti-CDC2 Mouse Monoclonal Antibody (CF488A) [clone: CDK1/873]

Supplier: Biotium

BNC880873-100 BNC880873-500
75967-420EA 325.12 CAD
75967-420 75967-422
Anti-CDC2 Mouse Monoclonal Antibody (CF488A) [clone: CDK1/873]
Antibodies
This antibody recognizes a 34 kDa protein, which is identified as cyclin dependent kinase 1 (cdk1) or p34cdc2 protein kinase. cdk1 / p34cdc2 plays a crucial role during cell division and is most active during mitosis. It is predominantly localized in the nucleus. It is a serine/threonine kinase, which is activated by cyclin, presumably by de-phosphorylation of tyrosine residues. Activated cdk1 / p34cdc2 performs specific functions during mitosis, including nuclear envelope breakdown and chromosome condensation.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®488A is a greenMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CDC2
Environmentally Preferable
Antigen name p34 / cdk1
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 488A
Clone CDK1/873
Reactivity Monkey,Human,Bovine,Mink
Host Mouse
Gene ID Unigene ID: 334562
Isotype IgG2a kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/ml
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR