Anti-RBP1 Mouse Monoclonal Antibody (Biotin) [clone: G4E4]

Supplier: Biotium

BNCB0855-100 BNCB0855-500
75973-656EA 325.12 CAD
75973-656 75973-658
Anti-RBP1 Mouse Monoclonal Antibody (Biotin) [clone: G4E4]
Antibodies
This MAb recognizes an epitope within the 74-182 C-terminal sequence (11kD peptide fragment) of human serum Cellular Retinol Binding Protein 1 (CRBP 1), a single-chain glycoprotein belonging to the superfamily of hydrophobic molecule transporter proteins, which is responsible for transport of retinol (vitamin A1) from the liver to peripheral target tissues, like the eye, where it mediates the cellular uptake. CRBP 1 is synthesized by hepatic parenchymal cells where it becomes bound to its ligand retinol and is then released into the circulation, where it binds further to the protein transthyretin, to form a transporting complex, which is big enough not to be lost by filtration through the kidney glomeruli. It is detected in nearly all tissues with higher expression in adult ovary, pancreas, pituitary gland, adrenal gland, and fetal liver.

Type: Primary
Antigen: RBP1
Clonality: Monoclonal
Clone: Clone G4E4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Chimpanzee, Goat, Human, Monkey, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR