Anti-CALD1 Mouse Monoclonal Antibody (CF568) [clone: CALD1/820 h-CALD]

Supplier: Biotium

BNC680821-100 BNC680821-500
75882-686EA 325.12 CAD
75882-686 75882-688
Anti-CALD1 Mouse Monoclonal Antibody (CF568) [clone: CALD1/820 h-CALD]
Antibodies
This antibody recognizes a protein of 150 kDa, which is identified as the high molecular weight variant of Caldesmon. Two closely related variants of human caldesmon have been identified which are different in their electrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150 kDa) is predominantly expressed in smooth muscle whereas l-caldesmon (70-80 kDa) is found in non- muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle. This MAb This antibody recognizes only the 150 kDa variant (h-caldesmon) in Western blots of human aortic media extracts and is unreactive with fibroblast extracts from cultivated human foreskin. Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: CALD1
Clonality: Monoclonal
Clone: Clone CALD1/820 h-CALD
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR