Anti-FUT4 Mouse Monoclonal Antibody [clone: FUT4/815]

Supplier: Biotium

BNUB0815-100 BNUB0815-500
75888-196EA 380.65 CAD
75888-196 75888-198
Anti-FUT4 Mouse Monoclonal Antibody [clone: FUT4/815]
Antibodies
CD15 plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. In addition, CD15 is expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkins disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.

Type: Primary
Antigen: FUT4
Clonality: Monoclonal
Clone: Clone FUT4/815
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR